2-(Difluoromethyl)-3,6-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

98460-500MG 98460-1G 98460-5G
77339-928EA 679.8 USD
77339-928 77359-330 77383-312
2-(Difluoromethyl)-3,6-difluorobenzoic acid ≥97%
2-(Difluoromethyl)-3,6-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD28788477
CAS Number: 1806363-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR