(5-Bromo-2-chloro-4-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

98386-500MG 98386-1G 98386-5G
77340-406EA 1027.94 USD
77340-406 77359-328 77383-310
(5-Bromo-2-chloro-4-fluorophenyl)(methyl)sulfane ≥97%
(5-Bromo-2-chloro-4-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅BrClFS
MW: 255.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34775621
CAS Number: 2169052-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR