(4-Bromo-2-fluoro-3-iodophenyl)(methyl)sulfane

Supplier: AOBChem USA

Warning

98350-500MG 98350-1G 98350-5G 98350-25G
77335-240EA 141.11 USD
77335-240 77344-956 77371-306 77391-732
(4-Bromo-2-fluoro-3-iodophenyl)(methyl)sulfane
(4-Bromo-2-fluoro-3-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅BrFIS
Storage Temperature: Ambient
MDL Number: MFCD34775608
CAS Number: 2710937-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR