(3-Chloro-4-methoxy-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

98152-500MG 98152-1G 98152-5G 98152-25G
77335-238EA 159.65 USD
77335-238 77345-756 77371-302 77391-730
(3-Chloro-4-methoxy-2-methylphenyl)(methyl)sulfane ≥97%
(3-Chloro-4-methoxy-2-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁ClOS
Storage Temperature: Ambient
MDL Number: MFCD22489462
CAS Number: 1822628-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR