2-Methoxy-4-methylphenyl acetate

Supplier: AOBChem USA

Warning

97907-1G 97907-5G 97907-25G
77341-378EA 82.4 USD
77341-378 77371-300 77383-306
2-Methoxy-4-methylphenyl acetate
2-Methoxy-4-methylphenyl acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD30214972
CAS Number: 879-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR