2-Bromo-6-methoxy-3-methylphenol ≥97%

Supplier: AOBChem USA

Warning

97892-500MG 97892-1G 97892-5G 97892-25G
77335-236EA 276.04 USD
77335-236 77359-326 77371-298 77391-728
2-Bromo-6-methoxy-3-methylphenol ≥97%
2-Bromo-6-methoxy-3-methylphenol

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD22414714
CAS Number: 129103-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR