(4-Bromo-2-fluoro-5-methoxyphenyl)(methyl)sulfane

Supplier: AOBChem USA

Warning

97873-500MG 97873-1G 97873-5G 97873-25G
77335-234EA 116.39 USD
77335-234 77343-476 77371-294 77383-302
(4-Bromo-2-fluoro-5-methoxyphenyl)(methyl)sulfane
(4-Bromo-2-fluoro-5-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈BrFOS
Storage Temperature: Ambient
MDL Number: MFCD34775439
CAS Number: 1403330-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR