4-(Benzyloxy)-2,6-dichloropyridine ≥97%

Supplier: AOBChem USA

Warning

97702-500MG 97702-1G 97702-5G 97702-25G
77335-232EA 191.58 USD
77335-232 77359-324 77371-292 77391-726
4-(Benzyloxy)-2,6-dichloropyridine ≥97%
4-(Benzyloxy)-2,6-dichloropyridine

Precaución: for Research Use Only

Formula: C₁₂H₉Cl₂NO
MW: 254.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09746245
CAS Number: 1262133-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR