4-Methyl-1-naphthol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Methylnaphthalen-1-ol

97317-500MG 97317-1G 97317-5G 97317-25G
77335-224EA 106.09 USD
77335-224 77343-142 77371-284 77383-294
4-Methyl-1-naphthol ≥97%
4-Methyl-1-naphthol

Precaución: for Research Use Only

Formula: C₁₁H₁₀O
MW: 158.196 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060344
CAS Number: 10240-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR