1-Bromo-3,5-dimethoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

96992-500MG 96992-1G 96992-5G 96992-25G
77335-222EA 276.04 USD
77335-222 77359-314 77371-282 77391-720
1-Bromo-3,5-dimethoxy-2-methylbenzene ≥97%
1-Bromo-3,5-dimethoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
MW: 231.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34774919
CAS Number: 62827-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR