(4-Fluoro-3-(trifluoromethoxy)phenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

96679-500MG 96679-1G 96679-5G
77335-612EA 360.5 USD
77335-612 77359-312 77383-288
(4-Fluoro-3-(trifluoromethoxy)phenyl)boronic acid ≥95%
(4-Fluoro-3-(trifluoromethoxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₄O₃
MW: 223.917 g/mol
Storage Temperature: Ambient
CAS Number: 881402-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR