2-Fluoro-3-(trifluoromethoxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

96669-1G 96669-5G 96669-25G
77340-970EA 64.89 USD
77340-970 77371-278 77383-286
2-Fluoro-3-(trifluoromethoxy)benzoic acid ≥95%
2-Fluoro-3-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₄O₃
MW: 224.109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12026477
CAS Number: 1159512-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR