4-Iodo-2-methoxy-6-(trifluoromethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

96565-500MG 96565-1G 96565-5G 96565-25G
77335-220EA 161.71 USD
77335-220 77346-044 77371-276 77391-718
4-Iodo-2-methoxy-6-(trifluoromethyl)pyridine ≥97%
4-Iodo-2-methoxy-6-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₅F₃INO
MW: 303.02 g/mol
Storage Temperature: Ambient
CAS Number: 1227581-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR