4-Bromo-3-(methoxymethyl)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

96436-500MG 96436-1G 96436-5G 96436-25G
77335-218EA 154.5 USD
77335-218 77345-558 77371-274 77391-716
4-Bromo-3-(methoxymethyl)benzoic acid ≥95%

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD26403783
CAS Number: 1141473-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR