4-Bromo-3-chloro-2-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

96402-500MG 96402-1G 96402-5G 96402-25G
77335-214EA 255.44 USD
77335-214 77359-310 77371-270 77391-714
4-Bromo-3-chloro-2-(methylthio)pyridine ≥95%
4-Bromo-3-chloro-2-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNS
MW: 238.533 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34774850
CAS Number: 2901103-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR