3-Chloro-2-(methylthio)isonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

96395-500MG 96395-1G 96395-5G 96395-25G
77335-212EA 164.8 USD
77335-212 77346-158 77371-268 77391-712
3-Chloro-2-(methylthio)isonicotinic acid ≥97%
3-Chloro-2-(methylthio)isonicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂S
MW: 203.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32116120
CAS Number: 1702581-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR