2-Bromo-4-chloro-6-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

96311-500MG 96311-1G 96311-5G 96311-25G
77335-210EA 267.8 USD
77335-210 77359-308 77371-266 77391-710
2-Bromo-4-chloro-6-(trifluoromethyl)phenol ≥97%
2-Bromo-4-chloro-6-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃O
MW: 275.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34774788
CAS Number: 1785285-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR