3-(Benzyloxy)-2-fluoro-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

96307-500MG 96307-1G 96307-5G 96307-25G
77335-208EA 124.63 USD
77335-208 77344-070 77371-264 77383-282
3-(Benzyloxy)-2-fluoro-4-(trifluoromethyl)benzaldehyde ≥97%
3-(Benzyloxy)-2-fluoro-4-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₀F₄O₂
MW: 298.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34774786
CAS Number: 2901102-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR