(4-iodo-2,5-dimethoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

96222-500MG 96222-1G 96222-5G
77335-982EA 371.83 USD
77335-982 77359-306 77383-280
(4-iodo-2,5-dimethoxyphenyl)(methyl)sulfane ≥97%
(4-iodo-2,5-dimethoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁IO₂S
Storage Temperature: Ambient
MDL Number: MFCD28793639
CAS Number: 1806354-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR