2,5-dimethoxy-4-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

96220-1G 96220-5G 96220-25G
77342-064EA 111.24 USD
77342-064 77371-260 77383-278
2,5-dimethoxy-4-(methylthio)benzaldehyde ≥97%
2,5-dimethoxy-4-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃S
MW: 212.265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863681
CAS Number: 61638-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR