(4-Bromo-2,5-dimethoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

96213-500MG 96213-1G 96213-5G 96213-25G
77335-204EA 267.8 USD
77335-204 77359-300 77371-258 77391-706
(4-Bromo-2,5-dimethoxyphenyl)(methyl)sulfane ≥97%
(4-Bromo-2,5-dimethoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂S
Storage Temperature: Ambient
MDL Number: MFCD34774761
CAS Number: 133042-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR