2,3-Dichloro-4-(methylthio)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

95770-1G 95770-5G 95770-25G
77342-136EA 114.33 USD
77342-136 77371-256 77383-270
2,3-Dichloro-4-(methylthio)benzaldehyde ≥95%
2,3-Dichloro-4-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆Cl₂OS
MW: 221.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34774701
CAS Number: 2901083-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR