4-Iodo-3-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

95724-500MG 95724-1G 95724-5G
77336-990EA 402.73 USD
77336-990 77359-296 77383-268
4-Iodo-3-methoxypyridine ≥95%
4-Iodo-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆INO
MW: 235.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24368704
CAS Number: 1331850-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR