4-Fluoro-3-methoxyphenyl acetate ≥97%

Supplier: AOBChem USA

Warning

95355-1G 95355-5G 95355-25G
77340-854EA 60.77 USD
77340-854 77371-254 77383-266
4-Fluoro-3-methoxyphenyl acetate ≥97%
4-Fluoro-3-methoxyphenyl acetate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD34774517
CAS Number: 2918921-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR