5-Chloro-6-methylpicolinic acid ≥95%

Supplier: AOBChem USA

Warning

95166-500MG 95166-1G 95166-5G
77339-804EA 645.81 USD
77339-804 77359-294 77383-262
5-Chloro-6-methylpicolinic acid ≥95%
5-Chloro-6-methylpicolinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257430
CAS Number: 137778-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR