5-Chloro-4-iodo-2,3-dimethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

95114-500MG 95114-1G 95114-5G 95114-25G
77335-202EA 120.51 USD
77335-202 77343-856 77371-250 77383-260
5-Chloro-4-iodo-2,3-dimethoxypyridine ≥97%
5-Chloro-4-iodo-2,3-dimethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇ClINO₂
Storage Temperature: Ambient
MDL Number: MFCD18803487
CAS Number: 1299607-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR