3-Bromo-2,4,6-trimethylbenzonitrile ≥97%

Supplier: AOBChem USA

Warning

95083-500MG 95083-1G 95083-5G
77338-088EA 462.47 USD
77338-088 77359-292 77383-258
3-Bromo-2,4,6-trimethylbenzonitrile ≥97%
3-Bromo-2,4,6-trimethylbenzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrN
MW: 224.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34774413
CAS Number: 1536674-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR