3,5-Difluorophenyl acetate ≥97%

Supplier: AOBChem USA

Warning

95076-500MG 95076-1G 95076-5G 95076-25G
77335-198EA 118.45 USD
77335-198 77343-578 77371-244 77383-254
3,5-Difluorophenyl acetate ≥97%
3,5-Difluorophenyl acetate

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂
MW: 172.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25977412
CAS Number: 315690-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR