(3-Chloro-2-ethoxy-5-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

94996-500MG 94996-1G 94996-5G 94996-25G
77335-196EA 95.79 USD
77335-196 77342-598 77371-242 77383-252
(3-Chloro-2-ethoxy-5-fluorophenyl)(methyl)sulfane ≥97%
(3-Chloro-2-ethoxy-5-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₀ClFOS
MW: 220.692 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34774378
CAS Number: 2710433-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR