3-Chloro-2-ethoxy-5-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

94992-500MG 94992-1G 94992-5G 94992-25G
77335-194EA 211.15 USD
77335-194 77359-288 77371-240 77391-702
3-Chloro-2-ethoxy-5-fluorobenzoic acid ≥97%
3-Chloro-2-ethoxy-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₈ClFO₃
MW: 218.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28627848
CAS Number: 1782054-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR