3-Bromo-2,4-difluoro-N-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

94970-500MG 94970-1G 94970-5G
77335-760EA 364.62 USD
77335-760 77359-286 77383-250
3-Bromo-2,4-difluoro-N-methoxy-N-methylbenzamide ≥97%
3-Bromo-2,4-difluoro-N-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₈BrF₂NO₂
MW: 280.066 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34774369
CAS Number: 2918910-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR