3-Chloro-2-fluoro-4-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

94402-500MG 94402-1G 94402-5G
77340-102EA 754.99 USD
77340-102 77359-282 77383-246
3-Chloro-2-fluoro-4-iodobenzaldehyde ≥97%
3-Chloro-2-fluoro-4-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClFIO
MW: 284.454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33437972
CAS Number: 2387241-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR