2-Methoxy-N,N-dimethylbenzamide ≥95%

Supplier: AOBChem USA

Warning

94236-1G 94236-5G 94236-25G
77341-706EA 100.94 USD
77341-706 77371-234 77383-244
2-Methoxy-N,N-dimethylbenzamide ≥95%
2-Methoxy-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01611815
CAS Number: 7291-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR