(5-Chloro-4-fluoro-2-isopropoxyphenyl)(methyl)sulfane ≥97%
Supplier: AOBChem USA
|
Warning
|
94158-500MG
94158-1G
94158-25G
94158-5G
77335-188EA
305.91
USD
77335-188
77359-278
77391-696
77383-242
(5-Chloro-4-fluoro-2-isopropoxyphenyl)(methyl)sulfane ≥97%
(5-Chloro-4-fluoro-2-isopropoxyphenyl)(methyl)sulfane
Precaución: for Research Use Only
Formula:
C₁₀H₁₂ClFOS Storage Temperature: Ambient |
MDL Number:
MFCD34773921 CAS Number: 2918944-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...