3-Chloro-2-fluoro-6-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

94125-500MG 94125-1G 94125-5G
77339-258EA 570.62 USD
77339-258 77359-276 77383-240
3-Chloro-2-fluoro-6-hydroxybenzoic acid ≥97%
3-Chloro-2-fluoro-6-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClFO₃
MW: 190.556 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28523057
CAS Number: 1780881-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR