3-Bromo-6-methyl-5-nitropyridin-2-amine ≥97%

Supplier: AOBChem USA

Warning

93906-500MG 93906-1G 93906-5G 93906-25G
77335-186EA 115.36 USD
77335-186 77343-406 77371-230 77383-238
3-Bromo-6-methyl-5-nitropyridin-2-amine ≥97%
3-Bromo-6-methyl-5-nitropyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₆BrN₃O₂
MW: 232.035 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00091868
CAS Number: 150935-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR