2,4-Difluoro-5-iodo-3-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

93697-500MG 93697-1G 93697-5G 93697-25G
77335-184EA 123.6 USD
77335-184 77344-040 77371-228 77383-236
2,4-Difluoro-5-iodo-3-isopropoxybenzaldehyde ≥97%
2,4-Difluoro-5-iodo-3-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉F₂IO₂
MW: 326.079 g/mol
Storage Temperature: Ambient
CAS Number: 2918959-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR