2-Bromo-1-(methoxymethyl)-4-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

93659-500MG 93659-1G 93659-5G 93659-25G
77335-178EA 217.33 USD
77335-178 77359-270 77371-222 77391-692
2-Bromo-1-(methoxymethyl)-4-(trifluoromethoxy)benzene ≥97%
2-Bromo-1-(methoxymethyl)-4-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₉H₈BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD34662249
CAS Number: 2901105-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR