3-(Benzyloxy)-2-methoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

93627-500MG 93627-1G 93627-5G 93627-25G
77335-176EA 198.79 USD
77335-176 77359-268 77371-220 77391-690
3-(Benzyloxy)-2-methoxy-5-methylbenzaldehyde ≥97%
3-(Benzyloxy)-2-methoxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MW: 256.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34773692
CAS Number: 2901103-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR