5-(Difluoromethoxy)-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

93526-500MG 93526-1G 93526-5G
77339-534EA 594.31 USD
77339-534 77359-266 77383-230
5-(Difluoromethoxy)-2-methylbenzaldehyde ≥97%
5-(Difluoromethoxy)-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34773607
CAS Number: 2384449-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR