2-Bromo-3-chloro-5-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

93502-500MG 93502-1G 93502-5G
77338-622EA 503.67 USD
77338-622 77359-264 77383-228
2-Bromo-3-chloro-5-fluorobenzoic acid ≥97%
2-Bromo-3-chloro-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrClFO₂
MW: 253.453 g/mol
Storage Temperature: Ambient
CAS Number: 1525442-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR