2-(Methylthio)-4-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

93492-500MG 93492-1G 93492-5G 93492-25G
77335-172EA 178.19 USD
77335-172 77346-632 77371-216 77391-686
2-(Methylthio)-4-(trifluoromethyl)benzaldehyde ≥97%
2-(Methylthio)-4-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇F₃OS
MW: 220.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28991158
CAS Number: 845740-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR