3,5-Dibromo-4'-methyl-[1,1'-biphenyl]-4-amine ≥97%
Supplier: AOBChem USA
|
Warning
|
93474-1G
93474-5G
93474-25G
77340-968EA
64.89
USD
77340-968
77371-214
77383-226
3,5-Dibromo-4'-methyl-[1,1'-biphenyl]-4-amine ≥97%
3,5-Dibromo-4'-methyl-[1,1'-biphenyl]-4-amine
Precaución: for Research Use Only
Formula:
C₁₃H₁₁Br₂N MW: 341.041 g/mol Storage Temperature: Ambient |
CAS Number:
2921730-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...