4-(4-(Trifluoromethyl)phenyl)tetrahydro-2H-pyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

93369-500MG 93369-1G 93369-5G 93369-25G
77335-164EA 272.95 USD
77335-164 77359-258 77371-204 77391-678
4-(4-(Trifluoromethyl)phenyl)tetrahydro-2H-pyran-4-ol ≥97%
4-(4-(Trifluoromethyl)phenyl)tetrahydro-2H-pyran-4-ol

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₃O₂
MW: 246.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18549617
CAS Number: 1411062-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR