4-Fluoro-3-(1-hydroxycyclohexyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

93352-500MG 93352-1G 93352-5G 93352-25G
77335-162EA 202.91 USD
77335-162 77359-256 77371-202 77391-676
4-Fluoro-3-(1-hydroxycyclohexyl)benzonitrile ≥97%
4-Fluoro-3-(1-hydroxycyclohexyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₄FNO
Storage Temperature: Ambient
MDL Number: MFCD34773506
CAS Number: 2918946-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR