1-(4-Chloro-2,3-difluorophenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

93340-500MG 93340-1G 93340-5G 93340-25G
77335-160EA 198.79 USD
77335-160 77359-254 77371-200 77391-674
1-(4-Chloro-2,3-difluorophenyl)cyclohexanol ≥95%
1-(4-Chloro-2,3-difluorophenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClF₂O
MW: 246.681 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34773499
CAS Number: 2918898-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR