2-Bromo-3,5-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

93311-1G 93311-5G 93311-25G
77341-586EA 93.73 USD
77341-586 77371-198 77383-222
2-Bromo-3,5-dimethoxybenzaldehyde ≥97%
2-Bromo-3,5-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD00797471
CAS Number: 85565-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR