1-(2,3-Difluorophenyl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

93287-500MG 93287-1G 93287-5G 93287-25G
77335-158EA 184.37 USD
77335-158 77347-348 77371-196 77391-672
1-(2,3-Difluorophenyl)cyclohexanol ≥97%
1-(2,3-Difluorophenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₂O
MW: 212.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19626870
CAS Number: 1339263-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR