1-(2,5-Difluorophenyl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

93144-500MG 93144-1G 93144-5G 93144-25G
77335-156EA 269.86 USD
77335-156 77359-252 77371-194 77391-670
1-(2,5-Difluorophenyl)-2-fluoroethanone ≥95%
1-(2,5-Difluorophenyl)-2-fluoroethanone

Precaución: for Research Use Only

Formula: C₈H₅F₃O
MW: 174.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34773425
CAS Number: 2293400-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR