1-Ethoxy-2-isopropyl-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

93082-500MG 93082-1G 93082-25G 93082-5G
77335-154EA 301.79 USD
77335-154 77359-248 77391-668 77383-218
1-Ethoxy-2-isopropyl-4-methylbenzene ≥97%
1-Ethoxy-2-isopropyl-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₈O
Storage Temperature: Ambient
MDL Number: MFCD34773387
CAS Number: 31574-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR